Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ West Nile Virus Capsid Protein C Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA5111982
Description
West Nile Virus Capsid Protein C Polyclonal Antibody for Western Blot
West Nile virus (WNV) is an infectious disease that first appeared in the United States in 1999. Infected mosquitoes spread the virus that causes it. People who get WNV usually have no symptoms or mild symptoms. The symptoms include a fever, headache, body aches, skin rash, and swollen lymph glands. WNV C prot.ein interaction with host lipid structures, essential for viral replication, is most relevant for future drug design.
Specifications
West Nile Virus Capsid Protein C | |
Polyclonal | |
Unconjugated | |
West Nile Virus | |
Full length West Nile virus Capsid protein C recombinant protein. | |
100 μL | |
Primary | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Western Blot | |
0.41 mg/mL | |
PBS with 1% BSA, 20% glycerol and 0.025% ProClin 300; pH 7 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
Virus | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction