Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
West Nile Virus Envelope Antibody (15), DyLight 755, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP306463IR
Description
West Nile Virus Envelope Monoclonal antibody specifically detects West Nile Virus Envelope in Virus samples. It is validated for Western BlotSpecifications
West Nile Virus Envelope | |
Monoclonal | |
DyLight 755 | |
50mM Sodium Borate | |
This antibody was produced from a hybridoma resulting from the fusion of a mouse myeloma with B cells obtained from a mouse immunized with purified, recombinant West Nile Virus (lineage 1, strain NY99) Envelope Protein (Domain III) (ABA62343.1; Gln586-Ala705). | |
0.1 mL | |
Primary | |
Store at 4°C in the dark. | |
IgG |
Western Blot | |
15 | |
Western Blot | |
Mouse | |
Protein A purified | |
RUO | |
Virus | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction