Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

West Nile Virus Envelope Antibody, Novus Biologicals™
SDP

Catalog No. NB10056744 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.025 mg
0.1 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB10056744 0.1 mg
NB10056744T 0.025 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. NB10056744 Supplier Novus Biologicals Supplier No. NB10056744
Only null left
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody has been used in 1 publication

West Nile Virus Envelope Polyclonal specifically detects West Nile Virus Envelope in Virus samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin).

Specifications

Antigen West Nile Virus Envelope
Applications Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Dilution Western Blot 1-3 ug/ml, Immunohistochemistry 1 ug/ml, Immunocytochemistry/Immunofluorescence reported in scientific literature (PMID 24920798), Immunohistochemistry-Paraffin 1 ug/ml
Formulation PBS with 0.05% Sodium Azide
Gene Accession No. P06935
Gene Alias West Nile Virus envelope glycoprotein E
Host Species Rabbit
Immunogen A synthetic peptide corresponding to 73-86 (CPTMGEAHNEKRAD) of West Nile virus envelope glycoprotein (NC_001563).
Purification Method Protein G purified
Quantity 0.1 mg
Regulatory Status RUO
Primary or Secondary Primary
Target Species Virus
Content And Storage Store at -20C. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.