Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
West Nile Virus Envelope Antibody (GT3029), Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP313574
This item is not returnable.
View return policy
Description
West Nile Virus Envelope Monoclonal antibody specifically detects West Nile Virus Envelope in Virus samples. It is validated for Western Blot, ImmunofluorescenceSpecifications
| West Nile Virus Envelope | |
| Monoclonal | |
| Unconjugated | |
| PBS | |
| Recombinant protein encompassing a sequence within the center region of West Nile Virus Envelope Protein (West Nile virus (strain NY99-IC)). The exact sequence is proprietary. | |
| 100 μg | |
| Immunology, Virology Bacteria and Parasites | |
| Virus | |
| Purified |
| Western Blot, Immunofluorescence | |
| GT3029 | |
| Western Blot 1:500-1:3000, Immunofluorescence 1:100-1:1000 | |
| Mouse | |
| Protein G purified | |
| RUO | |
| Primary | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. | |
| IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction