Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
West Nile Virus Envelope Antibody, mFluor Violet 610 SE, Novus Biologicals™

Rabbit Polyclonal Antibody
$555.00
Specifications
Antigen | West Nile Virus Envelope |
---|---|
Applications | ELISA |
Classification | Polyclonal |
Conjugate | mFluor Violet 610 SE |
Form | Purified |
Description
West Nile Virus Envelope Polyclonal antibody specifically detects West Nile Virus Envelope in Virus samples. It is validated for ELISASpecifications
West Nile Virus Envelope | |
Polyclonal | |
Purified | |
RUO | |
Virus | |
IgG | |
Peptide affinity purified |
ELISA | |
mFluor Violet 610 SE | |
Rabbit | |
Immunology, Virology Bacteria and Parasites | |
Antibody was raised against a synthetic peptide corresponding to 16 amino acids at the N terminus of the West Nile virus envelope protein. The immunogen is located within amino acids 350 - 400 of West Nile Virus Envelope Protein. Amino Acid Squence: CPTMGEAHNEKRADPA | |
Primary |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title