Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
West Nile Virus Envelope Antibody, PE, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP306464PE
Description
West Nile Virus Envelope Polyclonal antibody specifically detects West Nile Virus Envelope in Virus samples. It is validated for ELISASpecifications
West Nile Virus Envelope | |
Polyclonal | |
ELISA | |
Rabbit | |
Antigen and protein A Affinity-purified | |
RUO | |
Virus | |
Purified |
ELISA | |
PE | |
PBS | |
Produced in rabbits immunized with purified, recombinant WNV West Nile Virus Envelope (Accession#: ABA62343.1; Gln586-Ala705). | |
0.1 mL | |
Primary | |
Store at 4C in the dark. | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction