Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ West Nile Virus Envelope (Domain III) (lineage 1, strain NY99) Monoclonal Antibody (15)
GREENER_CHOICE

Catalog No. PIMA536036
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA536036 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIMA536036 Supplier Invitrogen™ Supplier No. MA536036
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

West Nile Virus Envelope (Domain III) (lineage 1, strain NY99) Monoclonal Antibody for Western Blot

West Nile Virus (WNV) is a member of the Flaviviridae, a plus-stranded virus family that includes St. Louis encephalitis virus, yellow fever virus, and Dengue virus. WNV was initially isolated in 1937 in the West Nile region of Uganda and has become prevalent in Africa, Asia, and Europe. It has rapidly spread across the United States with cases being observed in every continental state. Virus particles consist of a dense core made up of the core/capsid protein encapsulating the RNA genome surrounded by a membrane envelope embedded with envelope and matrix proteins which play a major role for WNV entry into target cells. The viral core protein is thought to contribute to the WNV-associated inflammation via apoptosis induced though the caspase-9 pathway as delivery of core gene delivery into the striatum of mouse brain and skeletal muscle resulted in cell death and inflammation.
TRUSTED_SUSTAINABILITY

Specifications

Antigen West Nile Virus Envelope (Domain III) (lineage 1, strain NY99)
Applications Western Blot
Classification Monoclonal
Clone 15
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with no preservative
Gene Alias West Nile envelope; West Nile Virus; WNV envelope
Host Species Mouse
Immunogen Recombinant West Nile Virus (WNV) (lineage 1, strain NY99) E/Envelope Protein (Domain III) Protein.
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Target Species Virus
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG2a
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.