Learn More
Invitrogen™ West Nile Virus Envelope Monoclonal Antibody (Q24Q)

Description
This antibody is specific for the envelope protein of West Nile Virus. It detects West Nile Virus and St. Louis Encephalitis, and does not react with Japanese Encephalitis, Yellow Fever, Dengue, Tick-Borne Encephalitis, Eastern Equine Encephalitis, Western Equine Encephalitis, Powassan, La Crosse Encephalitis or Venezuelan Equine Encephalitis. Purity is >90%.
Specifications
Specifications
| Antigen | West Nile Virus Envelope |
| Applications | ELISA, Immunocytochemistry |
| Classification | Monoclonal |
| Clone | Q24Q |
| Concentration | 100 μg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 0.1% sodium azide; pH 7.2 |
| Gene Alias | West Nile envelope; West Nile Virus; WNV envelope |
| Host Species | Mouse |
| Immunogen | West Nile Virus native protein. |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.