Learn More
Invitrogen™ West Nile Virus Glycoprotein M Polyclonal Antibody

Description
Suggested positive control: West Nile virus infected cell lysate. Immunogen sequence: GESTLANKKG AWLDSTKATR.
Specifications
Specifications
| Antigen | West Nile Virus Glycoprotein M |
| Applications | Immunohistochemistry (Paraffin), Western Blot |
| Classification | Polyclonal |
| Concentration | 1 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 0.05% sodium azide |
| Gene Alias | West Nile envelope; west nile virus glyco; WNV envelope; WNV Glycoprotein M |
| Host Species | Rabbit |
| Immunogen | A synthetic peptide corresponding to amino acid residues 8-27 of West Nile Virus MP. |
| Purification Method | Affinity chromatography |
| Show More |
Safety and Handling
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.