Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

West Nile Virus Mouse anti-Virus, Unconjugated, Clone: E16, MilliporeSigma™

Catalog No. MABF22342MI Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
100 μg
25 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MABF22342MI 25 μg
MABF22341MI 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. MABF22342MI Supplier MilliporeSigma Supplier No. MABF223425UG
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

West Nile Virus Monoclonal antibody specifically detects West Nile Virus in Virus samples. It is validated for Immunocytochemistry, Neutralizing, Western Blotting.
TRUSTED_SUSTAINABILITY

Specifications

Antigen West Nile Virus
Applications Immunocytochemistry, Neutralization, Western Blot
Classification Monoclonal
Clone E16
Conjugate Unconjugated
Gene Alias E protein;Envelope protein;Env
Gene Symbols E
Host Species Mouse
Immunogen West Nile Virus envelope protein.
Molecular Weight of Antigen ∼50 kDa observed; 31.88 kDa calculated. Uncharacterized bands may be observed in some lysate(s).
Purification Method Protein G Purified
Quantity 25 μg
Research Discipline Epigenetics and Nuclear Function
Primary or Secondary Primary
Test Specificity Clone E16 is a mouse monoclonal antibody that specifically detects envelope protein of West Nile Virus. It targets an epitope within the DIII lateral ridge region.
Target Species Virus
Content And Storage Stable for 1 year at -20°C from date of receipt.
Form Purified
Isotype IgG2b κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.