Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
West Nile Virus NS3 Protease Biotinylated Antibody, R&D Systems™

Goat Polyclonal Antibody has been used in 2 publications
Supplier: R&D Systems BAF2907
This item is not returnable.
View return policy
Description
wnvNS3 Protease Polyclonal specifically detects wnvNS3 Protease in Viral samples. It is validated for Western Blot.Specifications
wnvNS3 Protease | |
Polyclonal | |
Biotin | |
ABD85079 | |
Goat | |
Affinity Purified | |
RUO | |
912267 | |
Reconstitute at 0.2 mg/mL in sterile PBS. | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
Western Blot | |
LYOPH | |
Western Blot 0.1 ug/mL | |
flavivirus polyprotein gene | |
E. coli-derived recombinant wnvNS3 Protease, expressed as a fusion protein with cofactor NS2L Gly1506-Leu1689 Accession # ABD85079 | |
50 μg | |
Primary | |
Detects Viral wnvNS3 Protease in direct ELISAs and Western blots. | |
Virus | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction