Learn More
Carolina™ Wetting Solution
SDPSCI_ED
Catalog No. s37621
Click to view available options
Capacity (Metric):
0.4 L
3.7 L
There are Shipping Alerts for this Product Group Shipping Alert 1: Because this item is a living or perishable material a specified delivery date is required. Orders must be placed through Customer Service at 1-800-766-7000
There are Shipping Alerts for this Product Group Shipping Alert 1: Because this item is a living or perishable material a specified delivery date is required. Orders must be placed through Customer Service at 1-800-766-7000
For use as humectant and mold inhibitor for preserved specimens. Carolina™ Wetting Solution helps maintain moisture and reduce dehydration.
  • Helps keep tissue pliable and easy to work with, especially over extended period of time
  • Includes mold inhibitor
Specifications
Capacity (English) 1 gal.
Capacity (Metric) 3.7 L
For Use With (Application) Helps maintain moisture, reduce dehydration, and keep tissue pliable and easy to work with
Product Type Wetting solution
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.