Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ WFDC1 Polyclonal Antibody
Rabbit Polyclonal Antibody
$209.50 - $511.50
Specifications
Antigen | WFDC1 |
---|---|
Concentration | 500 μg/mL |
Content And Storage | -20°C |
Applications | Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry |
Classification | Polyclonal |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
PIPA5119507
|
Invitrogen™
PA5119507 |
200 μL |
Each for $511.50
|
|
|||||
PIPA5121497
|
Invitrogen™
PA5121497 |
20 μL |
Each for $209.50
|
|
|||||
Description
WFDC1 Polyclonal Antibody for Western Blot, ICC/IF, IHC (P)
This gene encodes a member of the WAP-type four disulfide core domain family. The WAP-type four-disulfide core domain, or WAP signature motif, contains eight cysteines forming four disulfide bonds at the core of the protein, and functions as a protease inhibitor in many family members. The encoded protein shares 81% amino acid identity with the rat ps20 protein, which was originally identified as a secreted growth inhibitor. This gene is mapped to chromosome 16q24, an area of frequent loss of heterozygosity in cancers, including prostate, breast and hepatocellular cancers and Wilms' tumor. Owing to its location and a possible growth inhibitory property of its gene product, this gene is suggested to be a tumor suppressor gene.Specifications
WFDC1 | |
-20°C | |
Polyclonal | |
Liquid | |
IgG | |
Mouse | |
WFDC1 | |
2310058A03Rik; Prostate stromal protein ps20; PS20; ps20 growth inhibitor; WAP four-disulfide core domain 1; WAP four-disulfide core domain 1 homolog; WAP four-disulfide core domain protein 1; Wfdc1 | |
WFDC1 | |
Primary | |
Antigen Affinity Chromatography |
500 μg/mL | |
Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
Unconjugated | |
Rabbit | |
RUO | |
PBS with 50% glycerol and 0.05% ProClin 300 | |
Q9ESH5 | |
67866 | |
Recombinant protein WAP Four Disulfide Core Domain Protein 1. The antigen corresponds to amino acid range 24-208 of the target protein. | |
Antibody |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title