Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ WFDC11 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA5144154
Description
WFDC11 Polyclonal Antibody for IHC (P), ELISA
WFDC11 is a gene that encodes a protein belonging to the WAP (whey acidic protein) four-disulfide core domain family. This family of proteins contains a conserved cysteine-rich domain that is thought to have a role in host defense. WFDC11 is expressed in the salivary gland, pancreas, and prostate gland, and has been shown to have a potential role in the regulation of cell proliferation and differentiation.
Specifications
WFDC11 | |
Polyclonal | |
Unconjugated | |
WFDC11 | |
WAP11 | |
Rabbit | |
Protein G | |
RUO | |
259239 | |
-20°C or -80°C if preferred | |
Liquid |
ELISA, Immunohistochemistry (Paraffin) | |
3.5 mg/mL | |
PBS with 50% glycerol and 0.03% Proclin 300; pH 7.4 | |
Q8NEX6 | |
WFDC11 | |
Recombinant Human Protein WFDC11 protein (26-87AA). | |
100 μg | |
Primary | |
Human | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction