Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

WFDC12 Polyclonal Antibody, Invitrogen™

Catalog No. PIPA523873
Change view
Click to view available options
Quantity:
400 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA523873 400 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA523873 Supplier Thermo Scientific Supplier No. PA523873

Rabbit Polyclonal Antibody

This gene encodes a member of the WAP-type four-disulfide core (WFDC) domain family. The WFDC domain, or WAP signature motif, contains eight cysteines forming four disulfide bonds at the core of the protein, and functions as a protease inhibitor. Most WFDC gene members are localized to chromosome 20q12-q13 in two clusters: centromeric and telomeric. This gene belongs to the centromeric cluster.
TRUSTED_SUSTAINABILITY

Specifications

Antigen WFDC12
Applications Flow Cytometry, Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 0.4 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.09% sodium azide
Gene WFDC12
Gene Accession No. Q8WWY7
Gene Alias C20orf122; dJ211D12.4; protease inhibitor WAP2; putative protease inhibitor WAP12; single WAP motif protein 2; SWAM2; UNQ544/PRO844; WAP four-disulfide core domain 12; WAP four-disulfide core domain protein 12; WAP2; WFDC12; Whey acidic protein 2
Gene Symbols WFDC12
Host Species Rabbit
Immunogen KLH conjugated synthetic peptide between 66-94 amino acids from the C-terminal region of human WFDC12
Purification Method Antigen affinity chromatography, Protein A
Quantity 400 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 128488
Target Species Human
Content And Storage -20° C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.