Learn More
WFDC2 Mouse anti-Human, Clone: OTI1G7, lyophilized, TrueMAB™

Mouse Monoclonal Antibody
Supplier: OriGene CF801093

Description
Reconstitute with PBS (pH 7.3) and recommend to perform another round of desalting process using Product No. 7KMWCO
This gene encodes a protein that is a member of the WFDC domain family. The WFDC domain, or WAP Signature motif, contains eight cysteines forming four disulfide bonds at the core of the protein, and functions as a protease inhibitor in many family members. This gene is expressed in pulmonary epithelial cells, and was also found to be expressed in some ovarian cancers. The encoded protein is a small secretory protein, which may be involved in sperm maturation.Specifications
WFDC2 | |
Monoclonal | |
Unconjugated | |
WFDC2 | |
EDDM4, HE4, WAP5, dJ461P17.6 | |
Mouse | |
Affinity Chromatography | |
RUO | |
10406 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Lyophilized |
Western Blot | |
OTI1G7 | |
PBS with 8% trehalose and no preservative; pH 7.3 | |
Q14508 | |
WFDC2 | |
Human recombit protein fragment corresponding to amino acids 31-124 of human WFDC2 produced in E.coli. | |
100 μg | |
Primary | |
Human | |
Antibody | |
IgG1 |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.