Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ WFDC2 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA547826
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA547826 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA547826 Supplier Invitrogen™ Supplier No. PA547826
Add to Cart
Add to Cart

Sheep Polyclonal Antibody

Reconstitute at 0.2 mg/mL in sterile PBS.

This gene encodes a protein that is a member of the WFDC domain family. The WFDC domain, or WAP Signature motif, contains eight cysteines forming four disulfide bonds at the core of the protein, and functions as a protease inhibitor in many family members. This gene is expressed in pulmonary epithelial cells, and was also found to be expressed in some ovarian cancers. The encoded protein is a small secretory protein, which may be involved in sperm maturation.
TRUSTED_SUSTAINABILITY

Specifications

Antigen WFDC2
Applications ELISA, Immunoprecipitation, Western Blot
Classification Polyclonal
Concentration 0.2 mg/mL
Conjugate Unconjugated
Formulation PBS with 5% trehalose and No Preservative
Gene WFDC2
Gene Accession No. Q14508
Gene Alias 1600023A02Rik; dJ461P17.6; EDDM4; epididymal protein 4; epididymal secretory protein 4; Epididymal secretory protein E4; epididymis-specific, whey-acidic protein type, four-disulfide core; HE4; Major epididymis-specific protein E4; MGC57529; putative protease inhibitor WAP5; RE4; WAP domain containing protein HE4-V4; WAP domain-containing protein HE4; WAP four-disulfide core domain 2; WAP four-disulfide core domain protein 2; WAP5; Wfdc2
Gene Symbols WFDC2
Host Species Sheep
Immunogen Mouse myeloma cell line NS0-derived recombinant human HE4/WFDC2 Thr28-Phe124.
Purification Method Antigen affinity chromatography
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 10406
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Lyophilized
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.