Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ WFDC2 Polyclonal Antibody

Goat Polyclonal Antibody
Supplier: Invitrogen™ PA519293
Description
This antibody is predicted to react with canine based on sequence homology. This antibody is tested in Peptide ELISA: antibody detection limit dilution 1,000.
This gene encodes a protein that is a member of the WFDC domain family. The WFDC domain, or WAP Signature motif, contains eight cysteines forming four disulfide bonds at the core of the protein, and functions as a protease inhibitor in many family members. This gene is expressed in pulmonary epithelial cells, and was also found to be expressed in some ovarian cancers. The encoded protein is a small secretory protein, which may be involved in sperm maturation.
Specifications
| WFDC2 | |
| Polyclonal | |
| Unconjugated | |
| WFDC2 | |
| 1600023A02Rik; dJ461P17.6; EDDM4; epididymal protein 4; epididymal secretory protein 4; Epididymal secretory protein E4; epididymis-specific, whey-acidic protein type, four-disulfide core; HE4; Major epididymis-specific protein E4; MGC57529; putative protease inhibitor WAP5; RE4; WAP domain containing protein HE4-V4; WAP domain-containing protein HE4; WAP four-disulfide core domain 2; WAP four-disulfide core domain protein 2; WAP5; Wfdc2 | |
| Goat | |
| Ammonium sulfate precipitation | |
| RUO | |
| 10406 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| Western Blot | |
| 0.5 mg/mL | |
| TBS with 0.5% BSA and 0.02% sodium azide; pH 7.3 | |
| Q14508 | |
| WFDC2 | |
| Synthetic peptide sequence (GAEKTGVCPELQADQ) corresponding to the internal amino acids of WFDC2 (aa 29-44). | |
| 100 μg | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction