Learn More
Invitrogen™ WFDC2 Recombinant Rabbit Monoclonal Antibody (6K2F5)

Description
Positive Samples: HeLa, HT-29, SKOV3, Mouse liver, Mouse kidney Immunogen sequence: CTQECVSDSE CADNLKCCSA GCATFCSLPN DKEGSCPQVN INFPQLGLCR DQCQVDSQCP GQMKCCRNGC GKVSCVTPNF.
Specifications
Specifications
| Antigen | WFDC2 |
| Applications | ELISA, Western Blot, Immunocytochemistry |
| Classification | Recombinant Monoclonal |
| Clone | 6K2F5 |
| Concentration | 1.5 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 50% glycerol, 0.05% BSA and 0.02% sodium azide; pH 7.3 |
| Gene | WFDC2 |
| Gene Accession No. | Q14508, Q9DAU7 |
| Gene Alias | 1600023A02Rik; dJ461P17.6; EDDM4; epididymal protein 4; epididymal secretory protein 4; Epididymal secretory protein E4; epididymis-specific, whey-acidic protein type, four-disulfide core; HE4; Major epididymis-specific protein E4; MGC57529; putative protease inhibitor WAP5; RE4; WAP domain containing protein HE4-V4; WAP domain-containing protein HE4; WAP four-disulfide core domain 2; WAP four-disulfide core domain protein 2; WAP5; Wfdc2 |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.