Learn More
Invitrogen™ WFDC3 Monoclonal Antibody (5C12)

Description
Peptide Sequence: MDENCQAGEK CCKSGCGRFC VPPVLPPKLT MNPNWTVRSD SELEIPVP.
Specifications
Specifications
| Antigen | WFDC3 |
| Applications | ELISA, Western Blot |
| Classification | Monoclonal |
| Clone | 5C12 |
| Conjugate | Unconjugated |
| Formulation | PBS with no preservative; pH 7.4 |
| Gene | WFDC3 |
| Gene Accession No. | Q8IUB2 |
| Gene Alias | dJ447F3.3; protease inhibitor WAP14; putative protease inhibitor WAP14; WAP four-disulfide core domain 3; WAP four-disulfide core domain protein 3; WAP14; WFDC3; whey acidic protein 14 |
| Gene Symbols | WFDC3 |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.