Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
WFDC5 Antibody (M47-P2D5), Janelia Fluor™ 549, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP250363JF549
Description
WFDC5 Monoclonal specifically detects WFDC5 in Human samples. It is validated for Western Blot, ELISA, Immunohistochemistry.Specifications
WFDC5 | |
Monoclonal | |
Janelia Fluor 549 | |
50mM Sodium Borate with 0.05% Sodium Azide | |
WFDC5 | |
Ovalbumin-conjugated synthetic peptide TERVHDGRPG | |
0.1 mL | |
Primary | |
Human | |
Purified |
Western Blot, ELISA, Immunohistochemistry | |
M47-P2D5 | |
Western Blot, ELISA, Immunohistochemistry | |
dJ211D12.5, PRG5protease inhibitor WAP1, Putative protease inhibitor WAP1, WAP four-disulfide core domain 5, WAP four-disulfide core domain protein 5, WAP1p53-responsive gene 5 protein | |
Mouse | |
Protein A purified | |
Cancer, Cellular Signaling | |
149708 | |
Store at 4°C in the dark. | |
IgG1 κ |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction