Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

WFDC5 Rabbit anti-Human, Polyclonal, Bioss
SDP

Catalog No. 501986889
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-198-6889 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-198-6889 Supplier Bioss Supplier No. BS5896R
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

WFDC5 is a putative acid-stable proteinase inhibitor.This gene encodes a member of the WAP-type four-disulfide core (WFDC) domain family. Most WFDC proteins contain only one WFDC domain, and this encoded protein contains two WFDC domains. The WFDC domain, or WAP signature motif, contains eight cysteines forming four disulfide bonds at the core of the protein, and functions as a protease inhibitor. Most WFDC gene members are localized to chromosome 20q12-q13 in two clusters: centromeric and telomeric. This gene belongs to the centromeric cluster.

Specifications

Antigen WFDC5
Applications Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 1 μg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.09% sodium azide; pH
Gene WFDC5
Gene Accession No. Q8TCV5
Gene Alias BC019734; dJ211D12.5; p53-responsive gene 5 protein; PRG5; protease inhibitor WAP1; Putative protease inhibitor WAP1; UNQ3036/PRO9842; WAP four-disulfide core domain 5; WAP four-disulfide core domain protein 5; WAP1; WFDC5
Gene Symbols WFDC5
Host Species Rabbit
Immunogen KLH conjugated synthetic peptide derived from human WFDC5.
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 149708
Target Species Human
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.