Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
WFDC8 Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP26260825UL
Description
WFDC8 Polyclonal antibody specifically detects WFDC8 in Human samples. It is validated for Immunohistochemistry, Immunohistochemistry (Paraffin)Specifications
WFDC8 | |
Polyclonal | |
Immunohistochemistry 1:50 - 1:200, Immunohistochemistry-Paraffin 1:50 - 1:200 | |
C20orf170, dJ461P17.1, WAP8, WFDC8 WAP four-disulfide core domain 8 | |
This antibody was developed against a recombinant protein corresponding to amino acids: KHKPGLCPKERLTCTTELPDSCNTDFDCKEYQKCCFFACQKKCMDPFQEPCMLPVRHGNCNHEAQRWHFDFKNYRCTPFKYRGCEGNA | |
25 μL | |
Primary | |
Human | |
Purified |
Immunohistochemistry, Immunohistochemistry (Paraffin) | |
Unconjugated | |
PBS (pH 7.2) and 40% Glycerol | |
Rabbit | |
Protein A purified | |
RUO | |
90199 | |
Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction