Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ WFDC8 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA583625
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA583625 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA583625 Supplier Invitrogen™ Supplier No. PA583625
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: KHKPGLCPKE RLTCTTELPD SCNTDFDCKE YQKCCFFACQ KKCMDPFQEP CMLPVRHGNC NHEAQRWHFD FKNYRCTPFK YRGCEGNA Highest antigen sequence indentity to the following orthologs: Rat - 48%, Mouse - 55%.

This gene encodes a member of the WAP-type four-disulfide core (WFDC) domain family. The WFDC domain, or WAP signature motif, contains eight cysteines forming four disulfide bonds at the core of the protein, and functions as a protease inhibitor. The encoded protein contains a Kunitz-inhibitor domain, in addition to three WFDC domains. Most WFDC genes are localized to chromosome 20q12-q13 in two clusters: centromeric and telomeric. This gene belongs to the telomeric cluster. Two alternatively spliced transcript variants have been found for this gene, and they encode the same protein. [provided by RefSeq, Jul 2008].
TRUSTED_SUSTAINABILITY

Specifications

Antigen WFDC8
Applications Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 0.05 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene WFDC8
Gene Accession No. Q8IUA0
Gene Alias C20orf170; dJ461P17.1; epididymis secretory protein Li 292; HEL-S-292; protease inhibitor WAP8; putative protease inhibitor WAP8; testicular secretory protein Li 68; WAP four-disulfide core domain 8; WAP four-disulfide core domain protein 8; WAP motif protein 1; WAP8; WFDC8
Gene Symbols WFDC8
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human WFDC8. Recombinant protein control fragment (Product #RP-99446).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 90199
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.