Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ WFS1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5116717
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5116717 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5116717 Supplier Invitrogen™ Supplier No. PA5116717
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Positive Samples: MCF7, HeLa, Mouse heart, Mouse brain Immunogen sequence: MDSNTAPLGP SCPQPPPAPQ PQARSRLNAT ASLEQERSER PRAPGPQAGP GPGVRDAAAP AEPQAQHTRS RERADGTGPT KGDMEIPFEE VLERAKAGDP KAQTEVGKHY LQLAGDTDEE LNSCTAVDWL VLAAKQGRRE AVKLLRRCLA DRRGITSENE REVRQLSSET DLERAVRKAA LVMYWKLNPK KKKQVAVAEL LENVGQVNEH DGGAQPGPVP KSLQKQRRML ERLVSSESKN YIALDDFVEI TKKYAKGVIP SSLFLQDDED DDELAGKSPE DLPLR.

WFS1 is a novel component of Wolfram syndrome, a rare form of juvenile diabetes. WFS1 plays an important role in maintaining homeostasis of the endoplasmic reticulum (ER) in the pancreas. It is normally up-regulated during insulin secretion, whereas inactivation of the protein can cause ER stress. Chronic ER stress is a major involvement in Wolfram syndrome.
TRUSTED_SUSTAINABILITY

Specifications

Antigen WFS1
Applications ELISA, Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 1.37 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.05% ProClin 300; pH 7.3
Gene WFS1
Gene Accession No. O76024, P56695
Gene Alias AI481085; CTRCT41; WFRS; WFS; WFS1; WFSL; Wolfram antibody; Wolfram syndrome 1 (wolframin); Wolfram syndrome 1 homolog; Wolfram syndrome 1 homolog (human); Wolfram syndrome 1 protein homolog; wolframin; wolframin ER transmembrane glycoprotein
Gene Symbols WFS1
Host Species Rabbit
Immunogen Recombinant fusion protein containing a sequence corresponding to amino acids 1-285 of human WFS1 (NP_0059962).
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 22393, 7466, 83725
Target Species Human, Mouse, Rat
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.