Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ WFS1 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA576065
Description
The antibody was affinity-purified from rabbit antiserum by affinity-chromatography using epitope-specific immunogen and the purity is > 95% (by SDS-PAGE).
WFS1 is a novel component of Wolfram syndrome, a rare form of juvenile diabetes. WFS1 plays an important role in maintaining homeostasis of the endoplasmic reticulum (ER) in the pancreas. It is normally up-regulated during insulin secretion, whereas inactivation of the protein can cause ER stress. Chronic ER stress is a major involvement in Wolfram syndrome.
Specifications
WFS1 | |
Polyclonal | |
Unconjugated | |
WFS1 | |
AI481085; CTRCT41; WFRS; WFS; WFS1; WFSL; Wolfram antibody; Wolfram syndrome 1 (wolframin); Wolfram syndrome 1 homolog; Wolfram syndrome 1 homolog (human); Wolfram syndrome 1 protein homolog; wolframin; wolframin ER transmembrane glycoprotein | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
22393, 7466, 83725 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Western Blot, Immunocytochemistry | |
1 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.2 | |
O76024, P56695 | |
WFS1 | |
Recombinant full length Human WFS1. | |
100 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction