Learn More
WFS1 Rabbit anti-Human, Unconjugated, Polyclonal, Proteintech

Rabbit Polyclonal Antibody
$518.60
Specifications
Antigen | WFS1 |
---|---|
Concentration | 467 μg/mL |
Applications | Western Blot, Immunohistochemistry, Immunofluorescence |
Classification | Polyclonal |
Conjugate | Unconjugated |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
50-223-8547
![]() ![]() |
Proteintech Group Inc
269951AP150UL |
150 μL |
Each of 1 for $518.60
|
|
|||||
Description
The WFS1 antibody from Proteintech is a rabbit polyclonal antibody generated with fusion protein of human WFS1. This antibody recognizes human, mouse, rat antigen. The WFS1 antibody has been validated for the following applications: IF, IHC, ELISA analysis.
Specifications
WFS1 | |
Western Blot, Immunohistochemistry, Immunofluorescence | |
Unconjugated | |
Human, Rat, Mouse | |
IgG |
467 μg/mL | |
Polyclonal | |
Rabbit | |
7466 | |
Primary |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.