Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tocris Bioscience™ WH-4-023
GREENER_CHOICE

Catalog No. 541310
Change view
Click to view available options
Quantity:
10 mg
50 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
54-131-0 10 mg
54-135-0 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. 54-131-0 Supplier Tocris Bioscience™ Supplier No. 5413/10
Add to Cart
Add to Cart
This item is not returnable. View return policy

Potent and selective Lck and Src inhibitor; also inhibits SIK

WH-4-023 is a potent and selective Lck and Src inhibitor (IC50 values are 2 and 6 nM respectively). Exhibits >300-fold selectivity against p38α and KDR. Also potently inhibits SIK (IC50 values are 10, 22 and 60 nM for SIK 1, 2 and 3 respectively) and displays selectivity over a range of closely related kinases. Supports self-renewal of naive hESCs in combination with PD 0325901, CHIR 99021 and SB 590885 .

Specifications

Quantity 10 mg
Product Type WH-4-023
Purity >98%
Molecular Weight (g/mol) 568.67
Inhibitors WH-4-023

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.