Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Edvotek™ What's In My Lunch? Quantitative Milk Allergy ELISA Experiment Kit

Shop All EDVOTEK Products


Click to view available options
Class Size:
10 Lab Groups
Description
Requires
Various food samples to be tested, distilled or deionized water, beakers or flasks, paper towels, 37˚C incubator, disposable lab gloves, safety goggles, automatic micropipettes (5-50μl, 100-1000μl) and tips, digital camera or cell phone with camera (optional), computers with internet, image analysis program, and graphing program (optional)
- For 10 Lab Groups
- Complete in 2 hours
- Level: Advanced
Specifications
Specifications
Grade | 9 to Undergraduate |
Includes | instructions, 10X Dilution Buffer, Whey Antigen, 10X PBST (Wash Buffer), Primary Antibody, Secondary Antibody, Aminosalicylic Acid, Hydrogen Peroxide, microtiter plates, snap-top microcentrifuge tubes, homogenization pestles with tubes, 15 and 50 ml conical tubes, transfer pipettes |
Content And Storage | Refrigerator Storage (Some components) |
Class Size | 10 Lab Groups |
Product Type | What's In My Lunch? Quantitative Milk Allergy ELISA Experiment Kit |
Description | What's In My Lunch? Quantitative Milk Allergy ELISA Experiment Kit |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction