Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Cytiva Whatman™ 0.8 μm Pore Size (ME 27) Mixed Cellulose Ester Membranes, White/Black Grid

Catalog No. p-4781233
Click to view available options
Filter Type:
Gridded Filter with Pad
Gridded Filter without Pad

Whatman™ (ME 27) Mixed Cellulose Ester Membranes, White/Black Grid are particularly effective in applications requiring higher flow rates and larger volume filtration. Supplied with clearly defined grid lines at regularly spaced intervals to assist in microbiological colony counting.

  • Material: Mixed Cellulose Ester
  • Material (Membrane): ME 27
  • Pore Size: 0.8 μm
  • Size (Metric) Grid: 3.1 mm
  • Color: White
  • Grid Color: Black
  • Autoclavable: Yes
  • Sterility: Sterile
  • Thickness (Metric): 140 μm
  • Diameter (Metric): 47 mm
  • Flow Rate: 2.8 sec./100 mL/12.5cm² (at Δp = 0.9 bar)
  • Gas Flow Rate: 11.6 sec./100 mL (at Δp = 3 mbar)
  • Bubble Point: 1.3 bar
  • For Use With (Application): Filtration, cytology, air monitoring, aqueous HPLC samples, biological assays
TRUSTED_SUSTAINABILITY

Specifications

Autoclavable Autoclavable
Bubble Point 1.3 bar
Color White
Filter Type Gridded Filter with Pad
Material MCE
Material (Membrane) ME 27
Pore Size 0.8 μm
Sterility Sterile
Flow Rate 2.8 sec./100 mL/12.5 cm2 (at Δp = 0.9 bar)
For Use With (Application) Filtration, cytology, air monitoring, aqueous HPLC samples, biological assays
Gas Flow Rate 11.6 sec./100 mL (at Δp = 3 mbar)
Grid Color Black
Quantity 100 Pk.
Size (Metric) Grid 3.1 mm
Thickness (English) 0.006 in.
Thickness (Metric) 140 μm
Diameter (English) 1.85 in.
Diameter (Metric) 47 mm
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.