Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Cytiva Whatman™ 0.8 μm Pore Size (ME 27) Mixed Cellulose Ester Membranes, Plain

Catalog No. 09927186 Shop All Cytiva Products
Change view
Click to view available options
Diameter (Metric):
25 mm
50 mm
100 mm
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Diameter (Metric)
09-927-186 25 mm
09-927-187 100 mm
09-927-256 50 mm
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 09-927-186 Supplier Cytiva Supplier No. 10400906
Add to Cart
Add to Cart

Whatman™ (ME 27) Mixed Cellulose Ester Membranes, Plain are particularly effective in applications requiring higher flow rates and larger volume filtration including clarification or sterilization of aqueous solutions, particulate analysis and removal, air monitoring, and microbial analysis.

  • Material: Mixed Cellulose Ester
  • Material (Membrane): ME 27
  • Filter Type: Plain
  • Pore Size: 0.8 μm
  • Autoclavable: Yes
  • Sterility: Non-sterile
  • Bubble Point: 1.3 bar
  • Flow Rate: 2.8 sec./100 mL/12.5 cm² (at Δp = 0.9 bar)
  • Gas Flow Rate: 11.6 sec./100 mL (at Δp = 3 mbar)
  • For Use With (Application): Filtration, cytology, air monitoring, aqueous HPLC samples, biological assays
TRUSTED_SUSTAINABILITY

Specifications

Autoclavable Autoclavable
Bubble Point 1.3 bar
Material MCE
Material (Membrane) ME 27
Pore Size 0.8 μm
Sterility Non-sterile
Flow Rate 2.8 sec./100 mL/12.5 cm⊃ (at Δp = 0.9 bar)
For Use With (Application) Filtration, cytology, air monitoring, aqueous HPLC samples, biological assays
Gas Flow Rate 11.6 sec./100 mL (at Δp = 3 mbar)
Grid Color Plain
Surface Type Plain
Diameter (English) 0.98 in.
Diameter (Metric) 25 mm
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.