Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Cytiva Whatman™ 3 μm Pore Size (ME 29) Mixed Cellulose Ester Membranes, Plain

Catalog No. p-4781236
Click to view available options
Diameter (Metric):
25 mm
47 mm
50 mm

Whatman™ (ME 29) Mixed Cellulose Ester Membranes, Plain are particularly effective in applications requiring higher flow rates and larger volume filtration including clarification or sterilization of aqueous solutions, particulate analysis and removal, air monitoring, and microbial analysis.

  • Material: Mixed Cellulose Ester
  • Material (Membrane): ME 29
  • Filter Type: Plain
  • Pore Size: 3 μm
  • Autoclavable: Yes
  • Sterility: Non-sterile
  • Bubble Point: 1.3 bar
  • Flow Rate: 1.2 sec./100 mL/12.5cm² (at Δp = 0.9 bar)
  • Gas Flow Rate: 6.7 sec./100 mL (at Δp = 3 mbar)
  • For Use With (Application): Filtration, cytology, air monitoring, aqueous HPLC samples, biological assays
TRUSTED_SUSTAINABILITY

Specifications

Autoclavable Autoclavable
Material MCE
Material (Membrane) ME 29
Pore Size 3 μm
Sterility Non-sterile
Flow Rate 1.2 sec./100 mL/12.5 cm² (at Δp = 0.9 bar)
For Use With (Application) Filtration, cytology, air monitoring, aqueous HPLC samples, biological assays
Gas Flow Rate 6.7 sec./100 mL (at Δp = 3 mbar)
Surface Type Plain
Diameter (English) 0.98 in.
Diameter (Metric) 25 mm
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.