Learn More
Description
- Fast flow rate
- Recovers and measures food contaminants
Specifications
Specifications
| Description | Grade 8 Ruled Filter Paper 45 mm 100/Pk |
| Format | Disc |
| Material | Cellulose |
| For Use With (Application) | For routine investigations of foreign substances in a variety of sample types. |
| Grade | 8 |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.