Learn More
Cytiva Whatman™ 850-DS 8-Channel Filter Plate
Click to view available options
Type:
8 Channel
Description
- Exclusively designed for use with the optional filter module on the Agilent 850-DS Dissolution Sampling Station
- Simplifies filter replacement between timepoints
- Reliable alignment of the liquid path increases productivity by reducing the risk of jamming, and reducing leaks that may occur with other dissolution sample preparation systems
- Save time and eliminate errors associated with manual sampling
- Automated processing of up to 8 samples simultaneously
- Readily available in a wide range of pore sizes and materials
Specifications
Specifications
For Use With (Equipment) | Agilent 850-DS Dissolution Sampling Station |
Material (Membrane) | Glass Microfiber |
For Use With (Application) | Automated Sample Preparation in Dissolution Testing |
Quantity | 50/Pk. |
Type | 8 Channel |