Learn More
Description
Binder free filters
The Whatman TCLP Filter is manufactured using a binder free borosilicate glass microfiber with a particle retention rating of 0.6μm to 0.8μm
- Resistant to acids and bases
- Available in six diameters, including sizes to fit Büchner funnels and zero-headspace extraction units
- Pretreatment with acid eliminates need to acid-wash filters in your lab, for reduced analysis time and reagent costs
Compliance
Conform to requirements of U.S. EPA Method 1311 Toxicity Characteristic Leaching Procedure (TCLP), including 0.6 to 0.8μm particle retention requirement.
Specifications
Specifications
| Description | GE’s Whatman TCLP test filters are acid-treated with low metal content. They support TCLP testing by providing accurate information about TCLP limits to ensure the best possible disposal plan can be put into action. |
| Format | Disc |
| Material | Borosilicate Glass |
| Particle Retention | 0.7 μm |
| For Use With (Application) | Required for volatile samples and use with a zero headspace extractor. |
| Quantity | 50 pieces |
| Shape | Circle |
| Thickness (Metric) | 420 μm |
| Type | TCLP Limits Test Filter |
| Waterflow Rate | 60 mL/min |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.