Learn More
Cytiva Whatman™ AS 300 Series Multiple Vacuum Filtration Apparatus
GSA_VA
Catalog No. 09927418 Shop All Cytiva Products
Change view
Click to view available options
Volume (Metric):
100 mL
500 mL
Includes:
Stainless Steel support screen filter, 3 place manifold
Stainless Steel support screen filter, 3 place manifold, rapid closure clamp
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Volume (Metric) Includes
09-927-418 500 mL Stainless Steel support screen filter, 3 place manifold, rapid closure clamp
09-927-304 500 mL Stainless Steel support screen filter, 3 place manifold
09-927-565 100 mL Stainless Steel support screen filter, 3 place manifold, rapid closure clamp
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 09-927-418 Supplier Cytiva Supplier No. 10445835
Add to Cart
Add to Cart
Request a Quote
AS300/3 Multiple Vacuum Filtration Apparatus, three-place vacuum filtration system, stainless steel 500 ml, 47/50 mm, support screen
The apparatus can be autoclaved and sterilized by dry heat at up to 180°C. Suitable only for vacuum operation. If flushing tubes are used, do not exceed 1.3 bar (300 mbar over-pressure).
This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.
TRUSTED_SUSTAINABILITY
Specifications
Autoclavable Autoclavable
Material Stainless Steel
Includes Stainless Steel support screen filter, 3 place manifold, rapid closure clamp
Series AS310/3
Type Vacuum Filtration System
Volume (Metric) 500 mL