Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Cytiva Whatman™ Binder-Free Glass Microfiber Filters, Grade GF/C

Catalog No. 09927362 Shop All Cytiva Products
Change view
Click to view available options
Diameter (Metric):
21 mm
24 mm
25 mm
32 mm
37 mm
42.5 mm
47 mm
50 mm
55 mm
70 mm
90 mm
100 mm
110 mm
125 mm
150 mm
Packaging:
Bulk
Individually Bagged
Quantity:
100 Pieces
400 Pieces
17 product options available for selection
Product selection table with 17 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Diameter (Metric) Packaging Quantity
09-927-362 100 mm Individually Bagged 100 Pieces
09-874-30 21 mm Bulk 100 Pieces
09-874-32 24 mm Bulk 100 Pieces
09-874-32A 25 mm Bulk 100 Pieces
09-927-923 25 mm Bulk 400 Pieces
09-806-105 32 mm Bulk 100 Pieces
09-874-32B 37 mm Bulk 100 Pieces
09-874-34 42.5 mm Bulk 100 Pieces
09-874-35 47 mm Bulk 100 Pieces
09-927-423 50 mm Bulk 100 Pieces
09-874-36 55 mm Bulk 100 Pieces
09-874-38 70 mm Bulk 100 Pieces
09-874-39 90 mm Bulk 100 Pieces
09-805-23A 100 mm Bulk 100 Pieces
09-874-36A 110 mm Bulk 100 Pieces
09-874-37 125 mm Bulk 100 Pieces
09-874-37A 150 mm Bulk 100 Pieces
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 17 options available.
17 missing translation for 'options'
Catalog No. 09-927-362 missing translation for 'mfr' Cytiva missing translation for 'supplierNo' 18229916
Add to Cart
Add to Cart

Combines fine particle retention with good flow rate. Whatman™ Binder-Free Glass Microfiber Filters, Grade GF/C are ideal for the collection of suspended solids in potable water, natural and industrial wastes.

Whatman glass microfiber filters without binder feature low resistance to fluid flow and high particle-loading capacity. These filters can be used at temperatures up to 500°C. The fine capillary structure of glass fiber filters can absorb large quantities of water, enabling spot tests and liquid scintillation counting methods.

Glass microfiber filters are mainly used in applications such as air or water monitoring and can be used as pre-filters for membranes.

  • Medium to fast, high loading
  • Airflow: 6.7s/100mL
  • Air Monitoring
  • Clarification of aqueous liquids containing low to medium levels of fine particulates
  • Cell harvesting
  • Liquid scintillation counting
  • Marine chemistry and biochemistry
  • Radioimmunoassay procedures
TRUSTED_SUSTAINABILITY

Specifications

Basis Weight 53 g/m2
Description GE’s Whatman filter paper Grade GF/C glass filters offer a good flow rate and loading capacity that support total suspended solids analysis.
Format Disc
Material Borosilicate Glass
Particle Retention 1.2 μm
Flow Rate 10.5 sec./100 mL
For Use With (Application) Widely used for cell harvesting, liquid scintillation counting and binding assays where more loading capacity is required.Highly suitable for use in applications involving air filtration and for gravimetric analysis of volatile materials where ignition is
Grade Grade GF/C
Packaging Individually Bagged
Quantity 100 Pieces
Shape Circle
Temperature (English) Max. 932°F
Temperature (Metric) Max 500°C
Thickness (English) 0.01 in.
Thickness (Metric) 260 μm
Type Filter, Individually Bagged 100 mm circle (100 pcs)
Waterflow Rate 105 mL/min
Diameter (English) 3.93 in.
Diameter (Metric) 100 mm
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.