Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Cytiva Whatman™ Carbon-Cap Disposable Activated Carbon Capsules

Catalog No. 0974437 Shop All Cytiva Products
Change view
Click to view available options
Filtration Area:
26000 m2
82,000 m2
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Filtration Area
09-744-37 82,000 m2
09-744-40 26000 m2
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. 09-744-37 Supplier Cytiva Supplier No. 67041500
Add to Cart
Add to Cart
This item is not returnable. View return policy

Filled with high-purity, high-efficiency, acid-washed, granular-activated carbon. Whatman™ Carbon Cap Disposable Activated Carbon Capsules are suitable for adsorption of organics from air and removal of color, organics and chlorine from water.

  • Remove organic odors, color, and colloids from water and air
  • Activated carbon plus a pleated HEPA filter media retains 99.97% of all particles 0.3μm
  • Maximum housing pressure: 4.1 bar (60psi.)
  • Carbon-Cap 75 offers 26,000m2 activated carbon surface area
  • Carbon-Cap 150 offers 82,000m2 filter area equivalent

Applications:

For vacuum pump outlet removal of smoke and oil.

TRUSTED_SUSTAINABILITY

Specifications

Description Carbon Cap 150, capsule filter with activated carbon (1 pc)
Filtration Area 82,000 m2
For Use With (Application) For water, chemical, and reagent purification
Housing Material Polypropylene
Max. Pressure 4.1 bar (60 psi)
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.