Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Cytiva Whatman™ Carbon-Cap Disposable Activated Carbon Capsules
GSA_VA

Catalog No. 0974440 Shop All Cytiva Products
Change view
Click to view available options
Filtration Area:
26000 m2
82,000 m2
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Filtration Area
09-744-40 26000 m2
09-744-37 82,000 m2
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 09-744-40 Supplier Cytiva Supplier No. 67047500
Add to Cart
Add to Cart

Filled with high-purity, high-efficiency, acid-washed, granular-activated carbon. Whatman™ Carbon Cap Disposable Activated Carbon Capsules are suitable for adsorption of organics from air and removal of color, organics and chlorine from water.

  • Remove organic odors, color, and colloids from water and air
  • Activated carbon plus a pleated HEPA filter media retains 99.97% of all particles 0.3μm
  • Maximum housing pressure: 4.1 bar (60psi.)
  • Carbon-Cap 75 offers 26,000m2 activated carbon surface area
  • Carbon-Cap 150 offers 82,000m2 filter area equivalent

Applications:

For vacuum pump outlet removal of smoke and oil.

This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.

TRUSTED_SUSTAINABILITY

Specifications

Description Carbon Cap 75, capsule filter with activated carbon (1 pc)
Disposable Single-use
Material Activated Carbon
Filtration Area 26000 m2
For Use With (Application) For water, chemical, and reagent purification
Housing Material Polypropylene
Max. Pressure 4.1 bar (60 psi)
Type Capsule
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.