Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Cytiva Whatman™ Cellulose Papers, Grade 597NF

Catalog No. 09920148 Shop All Cytiva Products
Change view
Click to view available options
Dimensions (L x W):
50 m x 300 mm
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Dimensions (L x W)
09-920-148 50 m x 300 mm
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 09-920-148 Supplier Cytiva Supplier No. 10320875
Add to Cart
Add to Cart
This item is not returnable. View return policy

For a wide variety of analytical routine applications in various industries

  • Standard grade filter
  • Used in qualitative analytical techniques to determine and identify materials
TRUSTED_SUSTAINABILITY

Specifications

Format Roll
Material Cellulose
Grade Grade 597 NF
Porosity Medium Fine
Type Qualitative Filter Paper
Dimensions (L x W) 50 m x 300 mm
Length (Metric) 50 m
Width (Metric) 300 mm
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.