Learn More
Cytiva Whatman™ Filtration Accessories
Catalog No. 09800941 Shop All Cytiva Products
Change view
Click to view available options
Diameter (Metric) Bottom:
25 mm
Catalog No. Diameter (Metric) Bottom
09-800-941 25 mm
1 options
Catalog No. 09-800-941 Supplier Cytiva Supplier No. 230600
Only null left
Add to Cart
Add to Cart
For use with membrane hardware where extra support is needed for improved flow rate and throughput.
The polyester drain disc is binder free and has a thickness of 100 μm. It provides a flat surface to eliminate filter tearing or rupturing. It is also used as a separator between membrane layers in serial stack filtration applications. This chemically inert support disc is available in a variety of diameters for use in a range of devices.
TRUSTED_SUSTAINABILITY
Specifications
For Use With (Equipment) Membrane hardware which needs extra support for improved flow rate and throughput
Material Polyester
Type Drain Disc
Diameter (Metric) Bottom 25 mm