Learn More
QIAGEN QIAcard FTA Indicating Mini
Catalog No. p-4903114
Click to view available options
Quantity:
100 Pk.
25 Pk.
For collection, stabilization, processing, transport and archiving of buccal nucleic acid samples. Indicating QIAcard FTA Mini with 2 sample areas per card.
  • QIAcard Indicating FTA Cards include a pink dye that turns white when a colorless sample, e.g. buccal cells or saliva is applied.
  • Stabilization and protection of nucleic acids at room temperature with for spot volumes of 125 μL.
  • Cell lysis on contact, denatures proteins and protects DNA from degradation.
Specifications
Description Collect, stabilize, process, transport, and archive buccal DNA samples with Indicating FTA Micro Cards.
Sample Type Buccal Cells, Saliva
For Use With (Application) Forensics, Transgenic identification, Transfusion medicine/HLA typing, Plasmid screening, Food and agriculture testing, Drug discovery, Genomics, STR analysis, Animal identification, Whole genome amplification,Molecular biology
No. of Samples 2
Product Type Indicating FTA Mini Card
Quantity 100 Pk.
Sample Volume 125 μL
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.