Learn More
GE Healthcare Whatman™ FTA™ Nucleic Acid Collection, Storage and Purification Cards: FTA Mini Available on GSA/VA Contract for Federal Government customers only.
Collect, store, and transport DNA at room temperature
Manufacturer:  GE Healthcare WB120056
 View more versions of this product
Catalog No. 09-923-343
Add to cart
Description
Description
Patented FTA technology simplifies the isolation, storage, and processing of nucleic acids from biological sources including whole blood, buccal cells, and plant material. Sample is applied to the FTA card and allowed to dry; the cells are lysed on contact. The released DNA is captured, stabilized, and protected from nucleases, oxidation, and UV damage.
  • DNA from blood stored at room temperature on FTA cards exhibits no loss in PCR efficiency after 22 years and counting
  • FTA cards are available in white and as pink indicating cards for colorless fluids
  • Pink indicating cards disclose where colorless samples have been spotted by changing to white spot
This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.
Specifications
Specifications
Collect, stabilize, process, transport, and archive buccal DNA samples with Whatman Indicating FTA Micro Cards.
Indicating FTA Mini Card, 2 sample areas per card
125 mL
Forensics. Transgenic identification. Transfusion medicine/HLA typing. Plasmid screening. Food and agriculture testing. Drug discovery. Genomics. STR analysis. Animal identification. Whole genome amplification Molecular biology.
125μL
Documents
Accessories