Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Cytiva Whatman™ Grade 1 Qualitative Filter Papers

Catalog No. 09920115 Shop All Cytiva Products
Change view
Click to view available options
Diameter (Metric):
45 mm
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Diameter (Metric)
09-920-115 45 mm
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 09-920-115 Supplier Cytiva Supplier No. 1001045
Add to Cart
Edge
Add to Cart

The most widely used filter paper for routine laboratory applications, qualitative analytical separations, and clarifying liquids. The standard grade cellulose filter papers for medium flow rates.

  • Used in qualitative analytical techniques to determine and identify materials
  • The most widely used filter paper for routine applications with medium retention and flow rate
  • Grade 1 covers a wide range of laboratory applications and is frequently used for clarifying liquids
  • Used in qualitative analytical separations for precipitates such as lead sulfate, calcium oxalate (hot), and calcium carbonate
  • Agriculture: Used for soil analysis and seed testing procedures
  • Food Industry: Used for numerous routine techniques to separate solid foodstuffs from associated liquid or extracting liquid. Widely used in education for teaching simple qualitative analytical separations
  • Air Pollution Monitoring: Using circles or rolls, atmospheric dust is collected from airflow and the stain-intensity measured photometrically
  • Gas Detection: Paper is impregnated with a chromogenic reagent and color formation quantified by optical reflectance
TRUSTED_SUSTAINABILITY

Specifications

Airflow Rate 13 s/100 mL/in2
Basis Weight 87 g/m2
Format Disc
Material Cellulose
Particle Retention 11 μm
Flow Rate 57 mL/min.
For Use With (Application) Routine Applications with Medium Retention and Flow Rate
Grade Grade 1 (Standard)
Quantity 100/Pk.
Thickness (Metric) 180 μm
Type Qualitative Filter Paper
Diameter (Metric) 45 mm
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.