Learn More
Description
- Medium-fast filter paper
- High wet strength
- Increased total loading capacity
- Maintained flow rate
Specifications
Specifications
| Description | Quantitative Filter Paper |
| For Use With (Application) | fit conical filter funnels in repetitive or multiple analyses |
| Grade | 1573-/12 |
| Quantity | 100/Pk. |
| Type | Prepleated |
| Diameter (Metric) | 320 mm |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.