Learn More
Description
- Absorbent paper strips for removal of excess liquids, e.g., from microscope specimens
Specifications
Specifications
| Description | 556 Drypads 37x100 mm 100/Pk |
| Material | Cellulose |
| For Use With (Application) | Absorbent paper strips for the removal of excess liquids, e.g., from microscope specimens. |
| Grade | Grade 556 |
| Quantity | 100 Pk. |
| Type | Drypad |
| Dimensions (L x W) | 37 x 100 mm |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.