Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Cytiva Whatman™ Grade 91 Wet Strengthened Qualitative Filter Papers - Sheets

Catalog No. p-4908605
Click to view available options
Dimensions (L x W):
58 x 58 cm
61 x 61 cm

High wet strength due to the addition of chemically stable resin

  • Wet strengthened grade
  • General purpose creped filter
  • For less critical routine analysis
  • Used to assay sucrose in cane sugar

TRUSTED_SUSTAINABILITY

Specifications

Description Grade 91 Qualitative Filter Paper Wet-Strengthened, sheet, 58 x 58 cm
Format Sheet
Material Cellulose
Pore Size 10 μm
Filtration Speed 70 sec.
For Use With (Application) A general purpose creped filter for less critical routine analysis. Widely used to assay sucrose in cane sugar and within pharmaceutical laboratories for routine filtration.
Grade Grade 91
Quantity 500 Pk.
Thickness (Metric) 205 μm
Dimensions (L x W) 58 x 58 cm
Length (Metric) 580 mm
Width (Metric) 580 mm
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.