Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Cytiva Whatman Grade GF/C Glass Microfiber Filters, Binder Free, Ready to Use

Catalog No. 09928050 Shop All Cytiva Products
Change view
Click to view available options
Diameter (Metric):
4.7 cm
7.0 cm
9.0 cm
Type:
Filter, pre-rinsed and dried
RTU Filter, pre-weighed
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Diameter (Metric) Type
09-928-050 4.7 cm RTU Filter, pre-weighed
09-928-051 7.0 cm RTU Filter, pre-weighed
09-928-053 4.7 cm Filter, pre-rinsed and dried
09-928-054 7.0 cm Filter, pre-rinsed and dried
09-928-055 9.0 cm Filter, pre-rinsed and dried
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Catalog No. 09-928-050 Supplier Cytiva Supplier No. 3822047
Add to Cart
Add to Cart

Grade GF/C glass filters offer fine particle retention, good flow rate and loading capacity, and support total suspended solids analysis.

  • Good flow rate and loading capacity
  • Fine nominal particle retention of 1.2μm
  • Binder free
  • Applications include total suspended solids analysis, industrial waste analysis and potable water
  • GF/C RTU pre-treats traditional GF/C filter with washing, drying, cooling (in a desiccator), and weighing steps aligned to EN 872
  • Certified weight loss of less than 0.017mg/cm2 after preparation
  • Each RTU filter is placed in an individually barcoded aluminum pan that notes the filter weight
TRUSTED_SUSTAINABILITY

Specifications

Airflow Rate 6.7s/100mL/in2
Description GE’s ready-to-use Whatman filter paper Grade GF/C glass filters offer a good flow rate and loading capacity that support total suspended solids analysis with a fine nominal particle retention of 1.2 μm.
Material Borosilicate Glass
Particle Retention 1.2 μm
Pore Size 1.2 μm
For Use With (Application) Widely used for cell harvesting, liquid scintillation counting and binding assays where more loading capacity is required.Highly suitable for use in applications involving air filtration and for gravimetric analysis of volatile materials where ignition is
Grade Grade GF/C
Quantity 100 Pieces
Temperature (Metric) Max 550°C
Thickness (Metric) 260 μm
Type RTU Filter, pre-weighed
Waterflow Rate 105 mL/min
Diameter (Metric) 4.7 cm
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.