Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
QIAGEN QIAcard FTA Elute Indicating Micro
Click to view available options
Quantity:
100 Pk.
25 Pk.
Description
- FTA Elute technology for convenient water based elution of nucleic acids.
- QIAcard Indicating FTA Cards include a pink dye that turns white when a colorless sample, e.g. buccal cells or saliva is applied.
- Stabilization and protection of nucleic acids at room temperature with for spot volumes of 125 μL.
- Cell lysis on contact, denatures proteins and protects DNA from degradation.
Specifications
Specifications
| Content And Storage | Room Temperature |
| Description | Collect, process, transport, and store colorless biological samples, such as buccal or saliva samples. With FTA Elute, proteins remain tightly bound while DNA is eluted from the matrix. |
| For Use With (Application) | Multiplex PCR, Sequencing after PCR amplification, SNP analysis, STR analysis, HLA typing, Whole genome amplification, Quantitative PCR, Biobanking, Pharmacogenomics, Genotyping, Genetic identification, Transgenic detection |
| Includes | Inert dye |
| Product Type | Indicating FTA Elute Micro Card |
| Quantity | 100 Pk. |
| Sample Volume | 40 μL |
| Test Time | 15 to 30 min |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction