Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Cytiva Whatman™ Mini-UniPrep™ Multicompressor and Accessories

Catalog No. 09928122 Shop All Cytiva Products
Change view
Click to view available options
Type:
Multicompressor
Tray
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Type
09-928-122 Multicompressor
09-928-123 Tray
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 09-928-122 Supplier Cytiva Supplier No. MUPMCPBC8
NLSU: Get 1 Consumable Product When You Buy 1
View all promotions
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Streamlines filtration, saves time, and reduces hand stress by enabling the filtration of eight Mini-UniPrep™ Vials at once. Trays to accomodate additional vials are sold separately.

  • Streamlines syringeless filtration by filtering up to eight filter vials simultaneously
  • Designed for Cytiva's Mini-UniPrep™ Standard Plastic Filter Vial
  • Mini-UniPrep™ Vials can be loaded directly into the autosampler
  • The multiprocessor includes one blue 8-position tray
  • Additional trays provide the added convenience of pre-loading up to eight Mini-UniPrep™ Filter Vials
TRUSTED_SUSTAINABILITY

Specifications

Color White, Purple
For Use With (Application) HPLC/UHPLC
Includes (1) 8-Position Tray for Standard Plastic Mini-UniPrep™ Filter Vials
Product Line Whatman
Quantity 1 Ea.
Type Multicompressor
Dimensions (L x W x H) 330 x 324 x 185 mm
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.